accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNN

CAS RN 6188-23-4

IUPAC Name: 6-bromoimidazo[1,2-a]pyridine
Synonyms: 6-bromoimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: FXPMFQUOGYGTAM-UHFFFAOYSA-N
SMILES: BrC1=CN2C=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results