accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61884-74-0

IUPAC Name: hydrogen (3S)-3-aminohexanedioic acid chloride
Synonyms: hydrogen (3S)-3-aminohexanedioic acid chloride
Molecular Weight (g/mol): 197.62
Molecular Formula: C6H12ClNO4
InChi Key: XMUKSNPNAOIQKU-WCCKRBBISA-N
SMILES: [H+].[Cl-].N[C@@H](CCC(O)=O)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)