accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OHONOO

CAS RN 619-17-0

IUPAC Name: 2-amino-4-nitrobenzoic acid
Synonyms: 2-amino-4-nitrobenzoic acid
Molecular Weight (g/mol): 182.14
Molecular Formula: C7H6N2O4
InChi Key: UEALKTCRMBVTFN-UHFFFAOYSA-N
SMILES: NC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)