accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-24-9

IUPAC Name: 3-nitrobenzonitrile
Synonyms: 3-cyano-1-nitrobenzene
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: RUSAWEHOGCWOPG-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(=C1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (3)
  • (4)
  • (1)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)