accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-42-1

IUPAC Name: methyl 4-bromobenzoate
Synonyms: methyl 4-bromobenzoate
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: CZNGTXVOZOWWKM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)