accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOI

CAS RN 619-44-3

IUPAC Name: methyl 4-iodobenzoate
Synonyms: methyl 4-iodobenzoate
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: DYUWQWMXZHDZOR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (5)
  • (4)
  • (4)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (5)