accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHO

CAS RN 619-64-7

IUPAC Name: 4-ethylbenzoic acid
Synonyms: 4-ethylbenzoic acid
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: ZQVKTHRQIXSMGY-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)