accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-66-9

IUPAC Name: 4-formylbenzoic acid
Synonyms: 4-formylbenzoic acid
Molecular Weight (g/mol): 150.13
Molecular Formula: C8H6O3
InChi Key: GOUHYARYYWKXHS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)