accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 619-80-7

IUPAC Name: 4-nitrobenzamide
Synonyms: 4-nitrobenzamide
Molecular Weight (g/mol): 166.14
Molecular Formula: C7H6N2O3
InChi Key: ZESWUEBPRPGMTP-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)