accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO

CAS RN 619-82-9

IUPAC Name: cyclohexane-1,4-dicarboxylic acid
Synonyms: cyclohexane-1,4-dicarboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: PXGZQGDTEZPERC-UHFFFAOYSA-N
SMILES: OC(=O)C1CCC(CC1)C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)