accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6190-65-4

IUPAC Name: 6-chloro-N2-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: desethylatrazine deethylatrazine desethyl atrazine 4-deethylatrazine deethylatrazin des-ethyl atrazine C6H10ClN5
Molecular Weight (g/mol): 187.63
Molecular Formula: C6H10ClN5
InChi Key: DFWFIQKMSFGDCQ-UHFFFAOYSA-N
SMILES: CC(C)NC1=NC(N)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results