accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNSBr

CAS RN 61929-24-6

IUPAC Name: 2-bromo-1,3,4-thiadiazole
Synonyms: 2-bromo-1,3,4-thiadiazole
Molecular Weight (g/mol): 165.01
Molecular Formula: C2HBrN2S
InChi Key: DQBCRVIBTFHJLM-UHFFFAOYSA-N
SMILES: BrC1=NN=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results