accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61970-08-9

IUPAC Name: 4-{6-[(1-hydroxy-3-methylbutan-2-yl)amino]imidazo[1,2-b]pyridazin-3-yl}benzonitrile
Synonyms: 4-{6-[(1-hydroxy-3-methylbutan-2-yl)amino]imidazo[1,2-b]pyridazin-3-yl}benzonitrile
Molecular Weight (g/mol): 321.38
Molecular Formula: C18H19N5O
InChi Key: CQZVJEKKNHOFNB-UHFFFAOYNA-N
SMILES: CC(C)C(CO)NC1=NN2C(C=C1)=NC=C2C1=CC=C(C=C1)C#N
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results