accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3(E)

CAS RN 6198-58-9

IUPAC Name: methyl (11E)-octadec-11-enoate
Synonyms: methyl (11E)-octadec-11-enoate
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: PVVODBCDJBGMJL-CMDGGOBGSA-N
SMILES: CCCCCC\C=C\CCCCCCCCCC(=O)OC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results