accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 61995-20-8

IUPAC Name: benzyl 3-oxopiperidine-1-carboxylate
Synonyms: benzyl 3-oxopiperidine-1-carboxylate
Molecular Weight (g/mol): 233.27
Molecular Formula: C13H15NO3
InChi Key: ALXLNFWWLXCXSK-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CCCC(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results