accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62-23-7

IUPAC Name: 4-nitrobenzoic acid
Synonyms: P-nitrobenzoic acid nitrodracylic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: OTLNPYWUJOZPPA-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)