accessibility menu, dialog, popup

UserName

Viewing profile as user:
CaCH3OOCH3OO

CAS RN 62-54-4

IUPAC Name: calcium diacetate
Synonyms: calcium acetate gray acetate calcium ethanoate brown acetate acetate of lime teltozan calac procalamine calcium diacetate sorbo-calcian
Molecular Weight (g/mol): 158.17
Molecular Formula: C4H6CaO4
InChi Key: VSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES: [Ca++].CC([O-])=O.CC([O-])=O
Grade 
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (5)
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (5)
  • (4)
  • (1)
  • (4)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)