accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHO

CAS RN 620-17-7

IUPAC Name: 3-ethylphenol
Synonyms: 3-ethylphenol M-ethylphenol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: HMNKTRSOROOSPP-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(O)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results