accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 620-19-9

IUPAC Name: 1-(chloromethyl)-3-methylbenzene
Synonyms: 1-(chloromethyl)-3-methylbenzene
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: LZBOHNCMCCSTJX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(CCl)=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)