accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 620-20-2

IUPAC Name: 1-chloro-3-(chloromethyl)benzene
Synonyms: 1-chloro-3-(chloromethyl)benzene
Molecular Weight (g/mol): 161.03
Molecular Formula: C7H6Cl2
InChi Key: DDGRAFHHXYIQQR-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(Cl)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results