accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPOCH3OH3C

CAS RN 620-42-8

IUPAC Name: tris(4-methylphenyl) phosphite
Synonyms: tris(4-methylphenyl) phosphite
Molecular Weight (g/mol): 352.37
Molecular Formula: C21H21O3P
InChi Key: FEVFLQDDNUQKRY-UHFFFAOYSA-N
SMILES: CC1=CC=C(OP(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results