accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3OOCH3

CAS RN 620-64-4

IUPAC Name: 1-(icosanoyloxy)-3-(nonadecanoyloxy)propan-2-yl henicosanoate
Synonyms: triarachidin
Molecular Weight (g/mol): 975.66
Molecular Formula: C63H122O6
InChi Key: ZQZYKZCGDCQGII-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCCCCC(=O)OC(COC(=O)CCCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)