accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 620-72-4

IUPAC Name: phenyl 2-bromoacetate
Synonyms: phenyl 2-bromoacetate
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: UEWYUCGVQMZMGY-UHFFFAOYSA-N
SMILES: BrCC(=O)OC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)