accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONO

CAS RN 62012-15-1

IUPAC Name: 1-(3-hydroxypropyl)pyrrolidin-2-one
Synonyms: 1-(3-hydroxypropyl)pyrrolidin-2-one
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: CVDGNRZPDAXOQO-UHFFFAOYSA-N
SMILES: OCCCN1CCCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results