accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62024-09-3

IUPAC Name: 2-[(2S)-6-azaniumyl-2-[(2S)-2-azaniumyl-3-(1H-imidazol-5-yl)propanamido]hexanamido]acetate
Synonyms: 2-[(2S)-6-ammonio-2-[(2S)-2-ammonio-3-(3H-imidazol-4-yl)propanamido]hexanamido]acetate
Molecular Weight (g/mol): 341.39
Molecular Formula: C14H25N6O4
InChi Key: XKIYNCLILDLGRS-QWRGUYRKSA-O
SMILES: [NH3+]CCCC[C@H](NC(=O)[C@@H]([NH3+])CC1=CN=CN1)C(=O)NCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)