CAS RN 6205-08-9
IUPAC Name:
5-amino-2-acetamidopentanoic acid
Synonyms:
5-amino-2-acetamidopentanoic acid
Molecular Weight (g/mol):
174.2
Molecular Formula:
C7H14N2O3
InChi Key:
JRLGPAXAGHMNOL-UHFFFAOYNA-N
SMILES:
CC(=O)NC(CCCN)C(O)=O
1
–
1
of
1
results
1
Promotions Available
N-Acetyl-L-ornithine, 95%
N-Acetyl-L-ornithine, 95%
CAS: 6205-08-9 Molecular Formula: C7H14N2O3 Molecular Weight (g/mol): 174.20 MDL Number: MFCD00065115 InChI Key: JRLGPAXAGHMNOL-UHFFFAOYNA-N PubChem CID: 439232 ChEBI: CHEBI:16543 IUPAC Name: (2S)-2-acetamido-5-aminopentanoic acid SMILES: CC(=O)NC(CCCN)C(=O)O