accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNONNNOOCH3OOH3COOCH3

CAS RN 6205-69-2

IUPAC Name: [3,4-bis(acetyloxy)-6-azido-5-acetamidooxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-6-azido-5-acetamidooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 372.33
Molecular Formula: C14H20N4O8
InChi Key: RMCFMPMNMQZHSF-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(OC(COC(C)=O)C(OC(C)=O)C1OC(C)=O)N=[N+]=[N-]
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results