accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62067-45-2

IUPAC Name: 4-(propan-2-yl)cyclohexane-1-carboxylic acid
Synonyms: 4-isopropylcyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: YRQKWRUZZCBSIG-UHFFFAOYSA-N
SMILES: CC(C)C1CCC(CC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results