accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOF(E/Z)(E/Z)

CAS RN 62085-74-9

IUPAC Name: 2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one
Synonyms: 2,6-bis[(4-fluorophenyl)methylidene]cyclohexan-1-one
Molecular Weight (g/mol): 310.34
Molecular Formula: C20H16F2O
InChi Key: STPNBQOTKFPYEG-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C2CCCC(=CC3=CC=C(F)C=C3)C2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results