accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHSNHH3C

CAS RN 621-01-2

IUPAC Name: 1,3-bis(4-methylphenyl)thiourea
Synonyms: 1,3-di-p-tolyl-2-thiourea
Molecular Weight (g/mol): 256.37
Molecular Formula: C15H16N2S
InChi Key: ULNVBRUIKLYGDF-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC(=S)NC2=CC=C(C)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results