accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-03-4

IUPAC Name: 2-cyano-N-phenylacetamide
Synonyms: acetamide, 2-cyano-N-phenyl-
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: XCTQPMCULSZKLT-UHFFFAOYSA-N
SMILES: O=C(CC#N)NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results