accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 621-07-8

IUPAC Name: N,N-dibenzylhydroxylamine
Synonyms: N,N-dibenzylhydroxylamine
Molecular Weight (g/mol): 213.28
Molecular Formula: C14H15NO
InChi Key: GXELTROTKVKZBQ-UHFFFAOYSA-N
SMILES: ON(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)