accessibility menu, dialog, popup

UserName

Viewing profile as user:
OS

CAS RN 621-08-9

IUPAC Name: (phenylmethanesulfinylmethyl)benzene
Synonyms: (phenylmethanesulfinylmethyl)benzene
Molecular Weight (g/mol): 230.33
Molecular Formula: C14H14OS
InChi Key: HTMQZWFSTJVJEQ-UHFFFAOYSA-N
SMILES: O=S(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results