accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-23-8

IUPAC Name: 1,3,5-trimethoxybenzene
Synonyms: 1,3,5-trimethoxybenzene 1,3,5-trimethyoxybenzene sym-trimethoxybenzene
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: LKUDPHPHKOZXCD-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(OC)=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (6)