accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-37-4

IUPAC Name: 2-(3-hydroxyphenyl)acetic acid
Synonyms: 3-hydroxyphenylacetic acid M-hydroxyphenylacetic acid metahydroxy phenylacetic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: FVMDYYGIDFPZAX-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=CC(O)=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (4)
  • (4)