accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-42-1

IUPAC Name: N-(3-hydroxyphenyl)acetamide
Synonyms: M-hydroxyacetanilide
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: QLNWXBAGRTUKKI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)