accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-70-5

IUPAC Name: 1,3-bis(hexanoyloxy)propan-2-yl hexanoate
Synonyms: tricaproin
Molecular Weight (g/mol): 386.53
Molecular Formula: C21H38O6
InChi Key: MAYCICSNZYXLHB-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCC(COC(=O)CCCCC)OC(=O)CCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results