accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 621-87-4

IUPAC Name: 1-phenoxypropan-2-one
Synonyms: 2-propanone, 1-phenoxy-
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: QWAVNXZAQASOML-UHFFFAOYSA-N
SMILES: CC(=O)COC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results