accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6211-16-1

IUPAC Name: dihydrogen (2S)-2,5-diaminopentanoic acid dichloride
Synonyms: dihydrogen L-ornithine dichloride
Molecular Weight (g/mol): 205.08
Molecular Formula: C5H14Cl2N2O2
InChi Key: HGBAVEGDXFHRQP-FHNDMYTFSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCC[C@H](N)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results