accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOONN

CAS RN 62124-77-0

IUPAC Name: ethyl 3-oxo-3-(pyrazin-2-yl)propanoate
Synonyms: ethyl 3-oxo-3-(pyrazin-2-yl)propanoate
Molecular Weight (g/mol): 194.19
Molecular Formula: C9H10N2O3
InChi Key: OPBDMDPBJKVMNL-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)C1=CN=CC=N1
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results