accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6213-19-0

IUPAC Name: 2-chloro-N,N-dimethyl-4-nitroaniline
Synonyms: 2-chloro-N,N-dimethyl-4-nitroaniline
Molecular Weight (g/mol): 200.62
Molecular Formula: C8H9ClN2O2
InChi Key: OZKAWTHGBGLZKC-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results