accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6214-44-4

IUPAC Name: (4-ethoxyphenyl)methanol
Synonyms: (4-ethoxyphenyl)methanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: UKFLLQIRBABMKF-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(CO)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)