accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62147-49-3

IUPAC Name: 1,3-dihydroxypropan-2-one
Synonyms: dihydroxyacetone vitadye ketochromin a,a'-dihydroxyacetone soleal aliphatic ketone summers brand of dihydroxyacetone Otan viticolor chromelin
Molecular Weight (g/mol): 90.08
Molecular Formula: C3H6O3
InChi Key: RXKJFZQQPQGTFL-UHFFFAOYSA-N
SMILES: OCC(=O)CO
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)