accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62150-45-2

IUPAC Name: 4-bromopyridine-2-carbonitrile
Synonyms: 4-bromopyridine-2-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: CZXDCTUSFIKLIJ-UHFFFAOYSA-N
SMILES: BrC1=CC(=NC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results