accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-03-7

IUPAC Name: 1,3-bis(phenylamino)thiourea
Synonyms: 1,3-bis(phenylamino)thiourea
Molecular Weight (g/mol): 258.34
Molecular Formula: C13H14N4S
InChi Key: BNSNUHPJRKTRNT-UHFFFAOYSA-N
SMILES: S=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results