accessibility menu, dialog, popup

UserName

Viewing profile as user:
O(E)(E)(E)(E)

CAS RN 622-21-9

IUPAC Name: (1E,3E,6E,8E)-1,9-diphenylnona-1,3,6,8-tetraen-5-one
Synonyms: (1E,3E,6E,8E)-1,9-diphenylnona-1,3,6,8-tetraen-5-one
Molecular Weight (g/mol): 286.37
Molecular Formula: C21H18O
InChi Key: RLJALOQFYHCJKG-FVRNMFRHSA-N
SMILES: O=C(\C=C\C=C\C1=CC=CC=C1)/C=C/C=C/C1=CC=CC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)