accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-22-0

OO

CAS RN 622-22-0

IUPAC Name: {[2-(benzyloxy)ethoxy]methyl}benzene
Synonyms: {[2-(benzyloxy)ethoxy]methyl}benzene
Molecular Weight (g/mol): 242.32
Molecular Formula: C16H18O2
InChi Key: FPFHYKOBYMYVAN-UHFFFAOYSA-N
SMILES: C(COCC1=CC=CC=C1)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results