accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 622-24-2

IUPAC Name: (2-chloroethyl)benzene
Synonyms: β-phenyl ethyl chloride
Molecular Weight (g/mol): 140.61
Molecular Formula: C8H9Cl
InChi Key: MNNZINNZIQVULG-UHFFFAOYSA-N
SMILES: ClCCC1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)