accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNO

CAS RN 622-40-2

IUPAC Name: 2-(morpholin-4-yl)ethan-1-ol
Synonyms: 4-morpholineethanol
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: KKFDCBRMNNSAAW-UHFFFAOYSA-N
SMILES: OCCN1CCOCC1
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (4)
  • (6)
  • (3)
  • (7)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)