accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-47-9

IUPAC Name: 2-(4-methylphenyl)acetate
Synonyms: tolylacetate
Molecular Weight (g/mol): 149.17
Molecular Formula: C9H9O2
InChi Key: GXXXUZIRGXYDFP-UHFFFAOYSA-M
SMILES: CC1=CC=C(CC([O-])=O)C=C1
Boiling Point 
  • (6)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (6)
  • (1)
  • (2)
  • (1)
  • (5)
  • (3)
  • (2)
  • (7)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)